Alfa Aesar™ Sodium hydrogen selenite, 98%

Manufacturer: Alfa Aesar™ A121730B

Quantity 1000g

 View more versions of this product

Catalog No. AAA121730B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-82-3
    Molecular Formula HNaO3Se
    Formula Weight 150.96
    MDL Number MFCD00003525
    UN Number UN2630
    Chemical Name or Material Sodium hydrogen selenite

    Specifications

    Formula Weight 150.96
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7782-82-3
    Quantity 1000g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
    Molecular Formula HNaO3Se
    MDL Number MFCD00003525
    UN Number UN2630
    Chemical Name or Material Sodium hydrogen selenite
    Assay 98%

    Description

    Applications
    Sodium hydrogen selenite is used as a culture media additive.

    Solubility
    Soluble in water.

    Notes
    Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit