Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrogen tartrate, 98+%, for analysis, Acros Organics

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
526-94-3
C4H5NaO6
172.068
MFCD00065393
NKAAEMMYHLFEFN-UHFFFAOYSA-M
sodium bitartrate,monosodium tartrate,sodium hydrogen tartrate,natriumtartrat german,sodium 3-carboxy-2,3-dihydroxypropanoate,monobasic sodium tartrate,monosodium l-+-tartrate,tartaric acid, monosodium salt,natrium hydrogen-2r,3r-tartrat,weinstein
23690454
sodium;2,3,4-trihydroxy-4-oxobutanoate
C(C(C(=O)[O-])O)(C(=O)O)O.[Na+]
526-94-3
C4H5NaO6
172.068
MFCD00065393
NKAAEMMYHLFEFN-UHFFFAOYSA-M
sodium bitartrate,monosodium tartrate,sodium hydrogen tartrate,natriumtartrat german,sodium 3-carboxy-2,3-dihydroxypropanoate,monobasic sodium tartrate,monosodium l-+-tartrate,tartaric acid, monosodium salt,natrium hydrogen-2r,3r-tartrat,weinstein
23690454
sodium;2,3,4-trihydroxy-4-oxobutanoate
C(C(C(=O)[O-])O)(C(=O)O)O.[Na+]
Merck Index 15, 8725
Arsenic (As) 1ppm max.
Formula Weight 172.07
Physical Form Powder
Percent Purity ≥99.0%
Grade Analysis
Calcium (Ca) 500ppm max.
Chloride 50ppm max.
Copper (Cu) 20ppm max.
Heavy Metals (as Pb) 20ppm max.
Insoluble in Water 0.03% max.
Iron (Fe) 20ppm max.
Lead (Pb) 20ppm max.
Nickel (Ni) 20ppm max.
Packaging Plastic bottle
Sulfate 100ppm max.
Color White
pH 3.3 to 3.6 (5% soln.)
Quantity 1kg
Chemical Name or Material Sodium hydrogen tartrate
Show More Show Less