Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrosulfite, ca. 85%, Tech., ACROS Organics™

Click to view available options
Quantity:
25g
1kg
2kg
10kg
Packaging:
Glass bottle
Plastic drum
7775-14-6
Na2O4S2
174.1
MFCD00011640
JVBXVOWTABLYPX-UHFFFAOYSA-L
sodium dithionite,sodium hydrosulfite,disodium dithionite,vatrolite,sodium sulfoxylate,sodium hydrosulphite,dithionous acid, disodium salt,blankit,burmol,hydros
24489
CHEBI:66870
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
7775-14-6
Na2O4S2
174.1
MFCD00011640
JVBXVOWTABLYPX-UHFFFAOYSA-L
sodium dithionite,sodium hydrosulfite,disodium dithionite,vatrolite,sodium sulfoxylate,sodium hydrosulphite,dithionous acid, disodium salt,blankit,burmol,hydros
24489
CHEBI:66870
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
Reduction reagent
Assay Percent Range ca. 85%
Linear Formula Na2S2O4
Merck Index 15, 8747
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 174.1
Physical Form Crystalline Powder
Percent Purity ca. 85%
Grade Technical
Packaging Plastic drum
Solubility Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
Color White
Melting Point >300.0°C
Quantity 10kg
Chemical Name or Material Sodium hydrosulfite, tech., ca. 85%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Self-heating; may catch fire.
Contact with acids liberates toxic gas.

GHS P Statement
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger

EINECSNumber : 231-890-0

TSCA : TSCA