Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.0100N, APHA for Sulfide, Ricca Chemicals

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 72952.5

Packaging Cubitainer
Quantity 10L

 View more versions of this product

Catalog No. 7295-25


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1310-73-2 (Sodium Hydroxide),7732-18-5 (Water, Deionized)
Sodium Hydroxide

Specifications

10L
0.0100N
APHA for Sulfide
1310-73-2 (Sodium Hydroxide),7732-18-5 (Water, Deionized)
Cubitainer
Sodium Hydroxide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit