Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Sodium hydroxide, 0.01N Standardized Solution
Click to view available options
Quantity:
1L
4L
20L
Specifications
Concentration | 0.01N |
Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
Formula Weight | 40 |
Physical Form | Liquid |
Odor | Odorless |
Quantity | 20L |
Chemical Name or Material | Sodium hydroxide, Standardized Solution |
Safety and Handling
EINECSNumber : 215-185-5
TSCA : Yes
Recommended Storage : Ambient temperatures