Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 0.0500 N (N/20), Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
CAS Min % | 98.0, 0.39 |
CAS Max % | 100.0, 0.41 |
Quantity | 500 mL |
Concentration or Composition (by Analyte or Components) | 0.0500N |
Packaging | Natural Poly Bottle |
Chemical Name or Material | Sodium Hydroxide |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction