Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.100 N Solution, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
6 x 1 L
12 x 1 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

S-340, 1310-73-2

Spectrum™ Sodium Hydroxide, 0.100 N Solution, APHA is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

CAS Max % 0.004,0.996
Quantity 4 x 4 L