Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.313 N (1.25%) Solution, AOAC for Crude Fiber, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
1310-73-2, 7732-18-5
1310-73-2, 7732-18-5
Concentration 0.313 N ± 0.003 N
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L

CAUTION: For Manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.