Sodium Hydroxide, 0.600N, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 74161

Packaging Plastic
Quantity 4L

 View more versions of this product

Catalog No. 7416-1

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Sodium Hydroxide

    Specifications

    Quantity 4L
    Concentration or Composition (by Analyte or Components) 0.600N
    Packaging Plastic
    Chemical Name or Material Sodium Hydroxide

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit