Sodium Hydroxide, 1.00 Normal, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 745055

Quantity 55 gal.

 View more versions of this product

Catalog No. 74-505-5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-73-2,7732-18-5
    Physical Form ear colorless liquid
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    Chemical Name or Material Sodium Hydroxide, 1.00 Normal

    Specifications

    Quantity 55 gal.
    Physical Form ear colorless liquid
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    CAS 1310-73-2,7732-18-5
    Chemical Name or Material Sodium Hydroxide, 1.00 Normal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit