Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1.0N Solution, Biotech Reagent, J.T.Baker™

Click to view available options
Quantity:
19L
Packaging:
Poly Bottle
Poly Drum

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Quantity 19L
Formula Weight 40
Packaging Poly Bottle
Specific Gravity 1L = 1.05kg
Chemical Name or Material Sodium Hydroxide, 1.0N Solution
Grade Biotech Reagent Grade