Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10.0 N Aqueous Solution, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

S-395, 1310-73-2

Spectrum™ Sodium Hydroxide, 10.0 N Aqueous Solution, APHA - As an APHA certified solution, this item meets the ASTM D1209 standards for use in water and waste water treatment facilities to determine the concentration of dissolved and particulate materials. Spectrum Chemical manufactured APHA grade products meet the toughest regulatory standards for quality and purity.

CAS Max % 0.3,0.7
Quantity 4 L