Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10N Solution, Biotech Reagent, J.T.Baker™

Click to view available options
Quantity:
19L
1L
4L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Quantity 4L
CAS 1310-73-2
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Packaging Poly Bottle
Specific Gravity 1L = 1.32kg
Chemical Name or Material Sodium Hydroxide, 10N Solution
Grade Biotech Reagent Grade
Show More Show Less