Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
19L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
Poly Bottle
Poly Pail
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Formula Weight 40
Packaging Poly Bottle
Storage Information Protect from air to avoid absorption of carbon dioxide; Material will freeze if stored below 41°F (5°C)
Specific Gravity 1L = 1.32kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Sodium Hydroxide, 10N Volumetric Solution