Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 12.5 N Solution, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
1310-73-2
1310-73-2
Concentration 12.5 N ± 0.5 N
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.