Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Formula Weight 40
Packaging Cubetainer
Storage Information Protect from air to avoid absorption of carbon dioxide
Specific Gravity 1L = 1.05kg
Chemical Name or Material Sodium Hydroxide, 1N Volumetric Solution
Grade BAKER ANALYZED™ Reagent