Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 2.50 N, Ricca Chemical
Click to view available options
Quantity:
25 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
25 mL HDPE Bottle
Cubitainer
Natural Poly Bottle
Poly Drum
Specifications
Color | Colorless |
Concentration | 2.499 to 2.501 N |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Quantity | 20 L |
IUPAC Name | sodium;hydroxide |
Molecular Weight (g/mol) | 39.997 |
Concentration or Composition (by Analyte or Components) | 2.50 N |
Packaging | Cubitainer |
Physical Form | Liquid |
Assay | Normal at 20°C |
Show More |
Safety and Handling
ShelfLife | 24 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction