Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 25% (w/v) (250 g/L), Ricca Chemical
SDP

Catalog Number 72731
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
1310-73-2, 7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium hydroxide
[OH-].[Na+]
1310-73-2, 7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium hydroxide
[OH-].[Na+]
Quantity 4 L
Concentration or Composition (by Analyte or Components) 25% (w/v) Aqueous Solution (250 g/L)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit