Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 4% (w/v) Aqueous Solution, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2,7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

S-414, 1310-73-2

Spectrum™ Sodium Hydroxide, 4 Percent (w/v) Aqueous Solution, (U.S.P. Test Solution) y raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

CAS Max % 0.04,0.96
Percent Purity 4% +/- 0.1%
Grade Chemical Solution
Packaging Poly Bottle
Quantity 4 L