Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% Solution, Biotech Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
19L
Packaging:
Poly Bottle
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Quantity 4L
Grade Biotech Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.53kg
Chemical Name or Material Sodium Hydroxide, 50% Solution