Sodium Hydroxide, 50% (w/w) Aqueous Solution, Analytical Reagent Grade, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 72912.5

Packaging Cubitainer™
Quantity 10L

 View more versions of this product

Catalog No. 7291-25

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-73-2,7732-18-5
    CAS Min % 49.0,49.0
    CAS Max % 51.0,51.0
    Chemical Name or Material Sodium Hydroxide
    Grade Analytical Reagent Grade

    Specifications

    Quantity 10L
    CAS 1310-73-2,7732-18-5
    Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution, 760g/L
    CAS Min % 49.0,49.0
    CAS Max % 51.0,51.0
    Packaging Cubitainer™
    Chemical Name or Material Sodium Hydroxide
    Grade Analytical Reagent Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit