Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) Aqueous Solution, Purified, Nitrogen Free, For Kjeldahl Nitrogen, Ricca Chemical

Click to view available options
Quantity:
4L
20L
Packaging:
Cubitainer™
Hard-Pack
Poly natural
1310-73-2, 7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium hydroxide
[OH-].[Na+]
1310-73-2, 7732-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium hydroxide
[OH-].[Na+]

For Analysis of Suitable For Kjeldahl Nitrogen Analysis

Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution (760 g/L)
Grade Purified
Packaging Poly natural
Quantity 4L
Chemical Name or Material Sodium Hydroxide Solution, Nitrogen Free