Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) Nitrogen-Free, Suitable for Kjeldahl Nitrogen Analysis, Reagents

Catalog No. 01336258
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-258 20 L Cubitainer
01-336-259 20 L -
01-336-257 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-258 Supplier Reagents Holdings Llc Supplier No. C731860020F
Only null left
Add to Cart
Add to Cart

Sodium Hydroxide, 50% (w/w) Nitrogen-Free, Suitable for Kjeldahl Nitrogen Analysis, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

Boiling Point 140°C
Color Colorless
CAS 1310-73-2, 7732-18-5
Physical Form Liquid
Quantity 20 L
Packaging Cubitainer
Percent Purity 50%
Grade Reagent
Chemical Name or Material Sodium Hydroxide, 50% (w/w) Nitrogen-Free, Suitable for Kjeldahl Nitrogen Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.