Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
Poly Bottle

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
CAS 1310-73-2
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Packaging Cubetainer
Storage Information Protect from air to avoid absorption of carbon dioxide
Specific Gravity 1L = 1.17kg
Chemical Name or Material Sodium Hydroxide, 5N Volumetric Solution
Grade BAKER ANALYZED™ Reagent
Show More Show Less