Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5383333
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 1 L
Formula Weight 40
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.17kg
Chemical Name or Material Sodium Hydroxide, 5N Volumetric Solution
Recommended Storage Protect from air to avoid absorption of carbon dioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.