Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
Poly Bottle

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Quantity 1L
Formula Weight 40
Packaging Narrow Mouth Poly Bottle
Storage Information Protect from air to avoid absorption of carbon dioxide
Specific Gravity 1L = 1.17kg
Chemical Name or Material Sodium Hydroxide, 5N Volumetric Solution
Grade BAKER ANALYZED™ Reagent