Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 98%, Acros Organics

Click to view available options
Quantity:
2.5kg
10kg
25kg
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Assay Percent Range 98%
Linear Formula NaOH
Formula Weight 40
Percent Purity 98%
Grade Pure
Packaging Plastic drum
Solubility Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
Melting Point 318.0°C
Boiling Point 1390.0°C
Quantity 10kg
Chemical Name or Material Sodium hydroxide
Show More Show Less