Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 98%, extra pure, pellets, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
25kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Aluminum (Al) 10ppm max.
Color Beige to Brown
Quantity 2.5kg
Calcium (Ca) 10ppm max.
Chloride 10ppm max.
Chlorine (Cl) 10ppm max.
Assay Percent Range 98%
Linear Formula NaOH
Density 2.13g/mL
Fieser 01,1083; 05,616; 07,336; 08,460
Merck Index 15, 8761
Heavy Metals (as Pb) 20ppm max.
Iron (Fe) 5ppm max.
Lead (Pb) 1ppm max.
Formula Weight 40
Physical Form Crystalline Powder
Percent Purity 98%
Grade Extra Pure
Nickel (Ni) 5ppm max.
Packaging Plastic Bottle
Phosphate 5ppm max.
Nitrogen Compounds 0.0005 % max. (as N)
Nitrogen (N) 0.0005% max.
Potassium (K) 200ppm max.
Silica 5ppm max.
Silicon Dioxide 5ppm max.
Silver (Ag) 0.5ppm max.
Sodium Carbonate 1.0 % max. (Na2CO3)
Solubility Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
Sulfate 10ppm max.
Melting Point 318.0°C
Boiling Point 1390.0°C
Chemical Name or Material Sodium hydroxide
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger