Alfa Aesar™ Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 1.0N

Manufacturer: Alfa Aesar™ 88619LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88619LS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-73-2
    Molecular Formula NaOH
    Formula Weight 40
    MDL Number MFCD00003548
    UN Number UN1824
    Chemical Name or Material Sodium hydroxide Acculute Standard Volumetric Solution, Final Concentration 1.0N

    Specifications

    Formula Weight 40
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003548
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    Quantity 6 units
    Physical Form Liquid
    Molecular Formula NaOH
    CAS 1310-73-2
    UN Number UN1824
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION
    Health Hazard 1 Danger
    Odor Odorless
    Chemical Name or Material Sodium hydroxide Acculute Standard Volumetric Solution, Final Concentration 1.0N

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit