Sodium Hydroxide Carbonate, 99.7-100.3%, ACS, Alfa Aesar™

For Use As a Source of CO2, in Making Sodium Salts, Baking Powder

Manufacturer: Alfa Aesar 1470736

Quantity 500g

 View more versions of this product

Catalog No. AA1470736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 144-55-8
    Physical Form Powder
    Assay Percent Range 99.7-100.3%
    Molecular Formula NaHCO3
    Formula Weight 84.01
    MDL Number MFCD00003528
    Synonym Sodium bicarbonate
    Chemical Name or Material Sodium hydrogen carbonate, For ACS analysis
    Grade ACS Grade

    Specifications

    Physical Form Powder
    Formula Weight 84.01
    Quantity 500g
    Color White
    Density 2.159g/cm3
    EINECS Number 205-633-8
    Odor Odorless
    Assay Percent Range 99.7-100.3%
    Molecular Formula NaHCO3
    MDL Number MFCD00003528
    CAS 144-55-8
    Synonym Sodium bicarbonate
    Health Hazard 1 H303: May be harmful if swallowed.
    Chemical Name or Material Sodium hydrogen carbonate, For ACS analysis
    Grade ACS Grade

    H303: May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit