Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, extra pure, 33wt.% solution in water, ACROS Organics™

Click to view available options
Quantity:
500mL
5L
25L
Packaging:
Plastic Bottle
Plastic drum
1310-73-2,7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2,7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Quantity 5L
Color Colorless
Chlorine (Cl) 50ppm max.
Linear Formula NaOH
Density 1.3600g/mL
Fieser 01,1083; 05,616; 07,336; 08,460
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 5ppm max.
Formula Weight 40
Physical Form Liquid
Percent Purity 32.5 to 33.5% (total base)
Grade Extra Pure
Packaging Plastic drum
Sodium Carbonate 0.5% (Na2CO3) max.
Solubility Solubility in water: soluble.
Specific Gravity 1.36
Melting Point 8.0°C
Boiling Point 120.0°C
Chemical Name or Material Sodium hydroxide
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger