Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, Flakes, Technical, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

S1308, 1310-73-2, NaOH

Spectrum™ Sodium Hydroxide, Flakes, Technical is a highly caustic metallic base combined with alkali salt. Commonly known as Caustic soda, this inorganic compound is water soluble, as well as in ethanol and methanol. The dissolution of solid Sodium Hydroxide in water creates a highly exothermic reaction (creates a lot of heat). One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Used in many industries, most notably in the manufacture of pulp, paper, textiles, drinking water, detergents and as a pharmaceutical excipient. Technical grade Sodum Hydroxide products are suitable for general laboratory use.

CAS Max % 1
UN Number UN1823
Grade Technical
Packaging Poly Bottle
Quantity 500 g