Sodium hydroxide (low chloride), ACS, 97.0% min, Alfa Aesar™

For Use in Chemical Manufacture, Rayon and Cellophane, As a Neutralizing Agent in Petroleum Refining

Manufacturer: Alfa Aesar 13455F6

Quantity 2 x 12.5kg

 View more versions of this product

Catalog No. AA13455F6

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1310-73-2
    Physical Form Pellets
    Assay Percent Range 97.0% min.
    Molecular Formula NaOH
    Formula Weight 40
    MDL Number MFCD00003548
    Chemical Name or Material Sodium hydroxide, (low chloride)
    Grade ACS Grade

    Specifications

    Physical Form Pellets
    Formula Weight 40
    Quantity 2 x 12.5kg
    Color White
    DOT Information Hazard Class: 8, Packing Group: II
    EINECS Number 215-185-5
    Odor Odorless
    Assay Percent Range 97.0% min.
    Molecular Formula NaOH
    Appearance White pellets
    MDL Number MFCD00003548
    CAS 1310-73-2
    Vapor Pressure 3.5 hPa (3mm Hg) at 800°C
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Sodium hydroxide, (low chloride)
    Grade ACS Grade

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit