Learn More
Sodium hydroxide monohydrate, 99.996% (metals basis), Thermo Scientific Chemicals
Description
Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Boiling Point | 1390°C |
Melting Point | 318.4°C |
Assay Percent Range | 99.996% (metals basis) |
Density | 2.13 |
Solubility Information | Soluble in water and methanol. |
Quantity | 100 g |
IUPAC Name | sodium hydrate hydroxide |
Formula Weight | 58.01 (40.00 Anhydrous) |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 215-185-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.