Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium hydroxide monohydrate, 99.996% (metals basis), Alfa Aesar™
Click to view available options
Quantity:
5g
25g
100g
Specifications
Appearance | White crystals |
Assay Percent Range | 99.996% (metals basis) |
Density | 2.13 g/cm3 at 20°C |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 58.01 (40.00 Anhydrous) |
Physical Form | Crystals |
Odor | Odorless |
Melting Point | 318.4°C |
Boiling Point | 1390°C |
Color | White |
Show More |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 215-185-5
TSCA : Yes
Recommended Storage : Ambient temperatures