Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, NF, 95-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

S2752, 1310-73-2, NaOH

Spectrum™ Sodium Hydroxide, NF
CAS Max % 1
Percent Purity 95 to 100.5%
Grade NF
Packaging Poly Bottle
Potassium (K) 0.005
Quantity 2.5 kg