Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Sodium hydroxide, pearl, 97%
Click to view available options
Quantity:
100g
500g
2500g
Description
Applications
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.
Solubility
Soluble in water, methanol, glycerol and ethanol. Insoluble in acetone and ether.
Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.
Solubility
Soluble in water, methanol, glycerol and ethanol. Insoluble in acetone and ether.
Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
Specifications
UN Number | UN1823 |
Merck Index | 14,8627 |
Solubility Information | Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether. |
Formula Weight | 40 |
Percent Purity | 97% |
Density | 2.13 |
Odor | Odorless |
Sensitivity | Air sensitive; Hygroscopic |
Boiling Point | 1390°C |
Melting Point | 318°C |
Show More |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE, SOLID
EINECSNumber : 215-185-5
RTECSNumber : WB4900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon