Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, pure, 1N Standard Solution, ACROS Organics™

Click to view available options
Quantity:
1L
5L
25L
Packaging:
Plastic Bottle
Plastic drum
1310-73-2,7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2,7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
Volumetric standard solution
Color Colorless
Quantity 5L
Concentration 90%98 to 1.002N (20°C)
Linear Formula NaOH
Density 1.0400g/mL
Fieser 01,1083; 05,616; 07,336; 08,460
Merck Index 15, 8761
Formula Weight 40
Physical Form Liquid
Packaging Plastic drum
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
Specific Gravity 1.04
Chemical Name or Material Sodium hydroxide
Show More Show Less

Danger

EINECSNumber : 215-185-5