Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 124260250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sodium hydroxide | |
1310-73-2, 7732-18-5 | |
NaOH | |
01,1083; 05,616; 07,336; 08,460 | |
1.0400g/mL | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
1.04 |
Colorless | |
HNaO | |
0.998 to 1.002N (20°C) | |
15, 8761 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
25 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Plastic Can | |
Liquid |
Safety and Handling
Danger
EINECSNumber : 215-185-5
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction