Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution, 50%(w/w), BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
19L
Packaging:
Jerrican (Poly Pail)
Poly Bottle

Chemical Identifiers

1310-73-2
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

High quality chemicals for laboratory and specialized industrial use

Quantity 19L
Formula Weight 40
Packaging Jerrican (Poly Pail)
Storage Information Protect from air to avoid absorption of carbon dioxide; IMPORTANT: Material will freeze if stored below 60°F (16°C)
Specific Gravity 1L = 1.53kg
Chemical Name or Material Sodium Hydroxide Solution
Assay 50% (w/w)
Grade BAKER ANALYZED™ Reagent