Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution, Biopharm Grade, 1.0 N, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
5 L
25 L
4 x 5 L
200 L
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

Spectrum™ Chemical cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Appearance of Solution Clear, colorless
Chloride 5.0 ppm
Assay Percent Range 3.65 to 4.05%
Concentration 0.9500 to 1.0500 N
Quantity 25 L
Heavy Metals 1 ppm
Grade Biopharm
Identification To pass test
Insoluble Subs. and Organic Matter To pass test
Iron (Fe) 0.2 ppm
Potassium (K) To pass test
Sodium Carbonate 0.002
Specific Gravity 1.039 to 1.043
Sulfate 10 ppm
Show More Show Less