Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide-Sulfide, for Kjeldahl Nitrogen Analysis using Mercury Catalyst, Ricca Chemical

Click to view available options
Quantity:
4L
20L
Packaging:
Cubitainer
Poly natural
1310-73-2, 7732-18-5, 39365-88-3
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2, 7732-18-5, 39365-88-3
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

For Analysis of Nitrogen (Kjeldahl) Using Mercrury Catalyst

Color Amber
Quantity 4L
CAS Min % 79.0, 18.53, 0.74
CAS Max % 82.0, 19.29, 0.77
Packaging Poly natural
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide-Sulfide