Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide on support, Granulated (0.8 to 1.6mm), For Elementary Analysis, MilliporeSigma™

Click to view available options
Quantity:
250g
1kg
Packaging:
Plastic bottle
1310-73-2, 14807-96-6
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2, 14807-96-6
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
For elementary analysis
Packaging Plastic bottle
Particle Size 0.8 to 1.6mm
Quantity 250g
UN Number UN 1823
Physical Form Solid
Chemical Name or Material Sodium Hydroxide on Support

H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II