Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium ingot, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number AA1034260
Click to view available options
Quantity:
450 g
5 x 450 g
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
5360545
CHEBI:26708
sodium
[Na]
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
5360545
CHEBI:26708
sodium
[Na]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray ingot
Assay Percent Range 99.8% (Metals basis)
Quantity 5 x 450 g
Grade ACS
Odor Odorless
Vapor Pressure 1.59 hPa (1mm Hg)
Form Ingot
Chemical Name or Material Sodium, Product meets ACS specifications
Hazard Category H260-H314-H335
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-132-9
RTECSNumber VY0686000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit