Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodate, 99+%, for analysis, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
7681-55-2
INaO3
197.89
MFCD00003533
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium iodate,natriumjodat german,iodic acid, sodium salt,sodium iodate naio3,iodic acid hio3 , sodium salt,sodium iodate naio3 pentahydrate,unii-u558pcs5z9,hsdb 749,iodic acid, sodium salt, pentahydrate,natriumjodat
23675764
CHEBI:81708
sodium;iodate
[O-]I(=O)=O.[Na+]
7681-55-2
INaO3
197.89
MFCD00003533
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium iodate,natriumjodat german,iodic acid, sodium salt,sodium iodate naio3,iodic acid hio3 , sodium salt,sodium iodate naio3 pentahydrate,unii-u558pcs5z9,hsdb 749,iodic acid, sodium salt, pentahydrate,natriumjodat
23675764
CHEBI:81708
sodium;iodate
[O-]I(=O)=O.[Na+]
Oxidation reagent
Additional Information bromide, bromate, chloride and chlorate (as Cl): < 0.02%
Assay Percent Range 99% min. (Iodometry)
Linear Formula NaIO3
Merck Index 15, 8764
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,insoluble in alcohol
Formula Weight 197.89
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Analysis
Copper (Cu) 5ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iodine (I) 10ppm max.
Iron (Fe) 10ppm max.
Lead (Pb) 5ppm max.
Loss on Drying 0.1% max. (130°C)
Nickel (Ni) 5ppm max.
Nitrogen (N) 0.003% max.
Packaging Plastic bottle
Potassium (K) 100ppm max.
Solubility Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol
Sulfate 50ppm max.
Color White
Melting Point 560.0°C
Quantity 2.5kg
Chemical Name or Material Sodium iodate, p.a.
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 231-672-5

RTECSNumber : NN1400000

TSCA : TSCA