Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide, 99.999%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
1g
10g
100g
Packaging:
Glass bottle
7681-82-5
INa
149.89
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide,ioduril,sodium monoiodide,soiodin,sodiumiodide,natrii iodidum,jodid sodny,iodure de sodium,jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
7681-82-5
INa
149.89
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide,ioduril,sodium monoiodide,soiodin,sodiumiodide,natrii iodidum,jodid sodny,iodure de sodium,jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula NaI
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
Merck Index 15, 8765
Solubility Information Solubility in water: 184g/100mL (25°C). Other solubilities: soluble in alcohol,glycerol and acetone
Formula Weight 149.89
Physical Form Granular Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 3.6600g/mL
Packaging Glass bottle
Solubility Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone
Specific Gravity 3.66
Total Trace Metal Impurities 0.001% max.
Boiling Point 1300.0°C
Color White
Melting Point 661.0°C
Quantity 1g
Chemical Name or Material Sodium iodide, Extra Pure
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

EINECSNumber : 231-679-3