Sodium iodide, ACS, 99.5% min, Alfa Aesar™

Used to prevent iodine deficiency. Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines, in baking ingredient conditioner. Also used for conversion of alkyl chlorides into alkyl iodides

Manufacturer: Alfa Aesar 4019622

Quantity 100g

 View more versions of this product

Catalog No. AA4019622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7681-82-5
    Physical Form Powder
    Assay Percent Range 99.5% min.
    Molecular Formula NaI
    Formula Weight 149.89
    MDL Number MFCD00003532
    Chemical Name or Material Sodium iodide
    Grade ACS Grade

    Specifications

    Physical Form Powder
    Boiling Point 1300° to 1304°C
    Quantity 100g
    Formula Weight 149.89
    TSCA Yes
    Health Hazard 2 P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a: Avoid breathing dust/fume/gas/mist/vapours/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Take off contaminated clothing and wash before reuse. Call a POISON CENTER or doctor/physician if you feel unwell. Specific treatment (see label). Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number WB6475000
    Melting Point 651°C
    Merck Index 14,8631
    Sensitivity Hygroscopic
    Solubility Soluble in water, alcohol, glycerol and acetone. Insoluble in ether.
    Color White
    Storage Note 1 Hygroscopic and incompatible with strong acids,oxidizing agents. light, air and moisture sensitive.
    Refractive Index 1.7745
    Density 3.67g/cm3 at 20°C
    EINECS Number 231-679-3
    Odor Odorless
    Assay Percent Range 99.5% min.
    Molecular Formula NaI
    Appearance White Powder
    MDL Number MFCD00003532
    CAS 7681-82-5
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Sodium iodide
    Grade ACS Grade

    Description

    Applications

    • Commonly used to prevent iodine deficiency Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines
    • Used in baking ingredient conditioner and as animal feed
    • Used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements

    P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a: Avoid breathing dust/fume/gas/mist/vapours/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Take off contaminated clothing and wash before reuse. Call a POISON CENTER or doctor/physician if you feel unwell. Specific treatment (see label). Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations.

    Hygroscopic and incompatible with strong acids,oxidizing agents. light, air and moisture sensitive.

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit