Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Iodide, Purified, Reagents

Catalog No. p-200021074
Click to view available options
Quantity:
125 g
500 g

Sodium Iodide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1304°C
Color White
Physical Form Solid
Quantity 125 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 3.67
Chemical Name or Material Sodium Iodide, Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.