Learn More
Sodium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1300°C to 1304°C |
Melting Point | 651°C |
Color | White |
Physical Form | Beads |
Quantity | 5 g |
Merck Index | 14,8631 |
Solubility Information | Very soluble in water and liquid ammonia. Freely soluble in acetone,formic acid,formamide,methanol,and acetonitrile. Soluble in N,N-dimethylformamide. |
Formula Weight | 149.89 |
Percent Purity | 99.999% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 231-679-3 |
RTECSNumber | WB6475000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.