Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Iodide, USP, 99-101.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7681-82-5
INa
149.89
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
[Na+].[I-]
7681-82-5
INa
149.89
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
[Na+].[I-]

SO175, 7681-82-5, NaI

Spectrum™ Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 99 to 101.5%
Iodate 4 ppm
Packaging Poly Pail
Water 0.02
Quantity 12 kg